Learn More
Liebherr™ LFK Series Laboratory Refrigerator
Product Code. 12527136 Shop All Liebherr Appliances Products
Change view
Click to view available options
Capacity (Metric) Refrigerator:
141 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Refrigerator Unit Size
12527136 141 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 12527136 Supplier Liebherr™ Supplier No. LFKUEX 1621
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Ensure maximum temperature stability. Liebherr™ LFK Series Laboratory Refrigerator is available in a range of sizes to meet every lab’s needs. Solid and glass door units feature automatic defrosting and rapid cooling. Electronic regulator includes audible and visual open door and high-temp alarms.
Advanced laboratory coolers
  • Electronic temperature regulation for maximum temperature stability
  • Digital display control panel with audible and visual open door and high-temperature alarms
  • Rapid cooling after door opening with eco-friendly refrigerant
  • ''Anti-freezing'' safety thermostat and automatic defrosting with collection tray
Constructed for durable, efficient operation
  • Constructed with high-density polyurethane insulation
  • ABS tanks with ATEX secure tank options
  • Epoxy treated steel bodywork, with solid and glass front doors
  • Range of sizes, from table-top to large capacity units
  • Reversible doors with key lock for safe storage of valuable goods
Specifications
Capacity (Metric) Refrigerator 141 L
Temperature Range (Refrigerator) 2°C to 12°C
Door Style Solid
Shelves 3
Defrost Automatic
Refrigerant R600A
Cabinet Material Epoxy-coated Steel
Insulation Polyurethane
Certifications/Compliance CE, ISO 14001, ISO 9001
Color White
Condenser Type Static
Includes ATEX secure tank 
Model LFKUEX1621
Depth (Metric) Exterior 61.5 cm
Depth (Metric) Interior 43.5 cm
Height (Metric) Exterior 82 cm
Height (Metric) Interior 66.7 cm
Width (Metric) Exterior 60 cm
Width (Metric) Interior 45.5 cm
Voltage 230 V
Frequency 50 Hz
Wattage 100 w
Shipping Weight (Metric) 39 kg