missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LGI2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76886-0.025mg
This item is not returnable.
View return policy
Description
LGI2 Polyclonal specifically detects LGI2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| LGI2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| FLJ10675, glioma inactivated 2, KIAA1916MGC126810, leucine-rich repeat LGI family, member 2, LGI1-like protein 2, MGC126808 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 55203 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 5 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| Q8K4Z0 | |
| LGI2 | |
| Antibody was raised against a 10 amino acid synthetic peptide from near the center of human LGI2. The immunogen is located within amino acids 290 - 340 of LGI2. Amino Acid Squence: IYKYDESWTK | |
| 0.025 mg | |
| Primary | |
| This LGI2 antibody is predicted to be specific to LGI2 and not recognize other LGI proteins.Three isoforms of LGI2 are known to exist; this LGI2 antibody will recognize only the longest. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction