missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LGMN Monoclonal Antibody (301417)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA561088

Product Code. 30535209

  • 429.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

This gene encodes a cysteine protease that has a strict specificity for hydrolysis of asparaginyl bonds. This enzyme may be involved in the processing of bacterial peptides and endogenous proteins for MHC class II presentation in the lysosomal/endosomal systems. Enzyme activation is triggered by acidic pH and appears to be autocatalytic. Protein expression occurs after monocytes differentiate into dendritic cells. A fully mature, active enzyme is produced following lipopolysaccharide expression in mature dendritic cells. Overexpression of this gene may be associated with the majority of solid tumor types. This gene has a pseudogene on chromosome 13. Several alternatively spliced transcript variants have been described, but the biological validity of only two has been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LGMN
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
O89017
LGMN
Mouse myeloma cell line NS0-derived recombinant mouse Legumain/Asparaginyl Endopeptidase, Val18-Tyr435.
100 μg
Primary
Mouse
Antibody
IgG2a
Western Blot
301417
Unconjugated
LGMN
AEP; AI746452; Asparaginyl endopeptidase; AU022324; cb173; cb460; cysteine protease 1; legumain; Lgmn; LGMN1; preprolegumain; protease, cysteine 1; protease, cysteine, 1; protease, cysteine, 1 (legumain); PRSC1; PRSC1EC 3.4.22.34; sb:cb173; wu:fa98g05; wu:fj92g08; zgc:76953
Rat
Protein A/G
RUO
19141
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.