Learn More
Lgr5/GPR49 Antibody (CL13579) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24571G
Product Code. 30307177
  • 737.00€ / 0.10mL
Description
Lgr5/GPR49 Monoclonal antibody specifically detects Lgr5/GPR49 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lgr5/GPR49
Monoclonal
DyLight 488
FEX, G protein-coupled receptor 49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38
This antibody was developed using a synthetic peptide derived from O75473, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, GPCR, Neuroscience, Signal Transduction, Wnt Signaling Pathway
8549
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL13579
50mM Sodium Borate
Mouse
Protein A purified