missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lgr6 Antibody (15G3), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00€ - 533.00€
Specifications
| Antigen | Lgr6 |
|---|---|
| Clone | 15G3 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30038491
|
Novus Biologicals
NBP3-26169-50ul |
50 μL |
343.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30029330
|
Novus Biologicals
NBP3-26169-100ul |
100 μL |
533.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lgr6 Monoclonal antibody specifically detects Lgr6 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
| Lgr6 | |
| Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ14471, gonadotropin receptor, GPCR, leucine-rich repeat containing G protein-coupled receptor 6, leucine-rich repeat-containing G protein-coupled receptor 6, leucine-rich repeat-containing G-protein coupled receptor 6, VTS20631 | |
| A synthesized peptide derived from Human Lgr6 [UniProt Q9HBX8] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 15G3 | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 59352 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title