Learn More
LHPP Mouse anti-Mouse, Unconjugated, Clone: 2A2A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66988-1-IG-20UL
Description
The LHPP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LHPP. This antibody recognizes mouse, rat, pig antigen. The LHPP antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| LHPP | |
| Monoclonal | |
| 913 μg/mL | |
| Mouse | |
| Primary | |
| Mouse, Rat, Pig |
| Western Blot, Immunofluorescence, ELISA | |
| 2A2A4 | |
| Unconjugated | |
| 20 μL | |
| 64077 | |
| IgG1 |