missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LHX2 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA518945
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator. The protein can recapitulate or rescue phenotypes in Drosophila caused by a related protein, suggesting conservation of function during evolution.
Specifications
| LHX2 | |
| Polyclonal | |
| Unconjugated | |
| Lhx2 | |
| ap; apterous; hLhx2; Homeobox protein LH-2; LH2; Lh-2; LH2A; LHX2; LIM homeo box protein 2; LIM homeobox 2; LIM homeobox protein 2; LIM HOX gene 2; LIM/homeobox protein Lhx2; Lim2; MGC138390 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 9355 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P50458 | |
| Lhx2 | |
| Synthetic peptide sequence (NLEGHEPHSPSQ) corresponding to the C-terminus amino acids of LHX2 (aa 388-400). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction