missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHX5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21567-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and be involved in the control of differentiation and development of the forebrain. In mice, this protein is essential for the regulation of precursor cell proliferation and the control of neuronal differentiation and migration during hippocampal development. This protein is involved in learning and motor functions in adult mice.Specifications
| LHX5 | |
| Polyclonal | |
| Unconjugated | |
| LHX5 | |
| LHX5, LIM homeobox 5, LIM homeobox protein 5, LIM/homeobox protein Lhx5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64211 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H2C1 | |
| LHX5 | |
| LHX5 Fusion Protein Ag16081 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction