Learn More
BOLA Liebig Condensers Transparent
Product Code. 10247351
Change view
Click to view available options
Length (Metric) Jacket:
450 mm
600 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Jacket unitSize
10247351 450 mm Each
10247401 600 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10247351 Supplier BOLA Supplier No. B29104
Add to basket
Add to basket
Liebig Condensers Transparent PTFE, with SAFE LAB-system
  • Thin-walled cooling tube made of translucent and gastight PFA
  • cooling jacket made of borosilicate glass with hose connectors
  • made of PP and nuts for connection of cooling water
  • ground joint cone and socket size 29 made of PTFE.
  • The distillate is only exposed to PFA/PTFE. Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.
Specifications
Certifications/Compliance FDA
Length (Metric) Jacket 450 mm