Learn More
Invitrogen™ LIFR Recombinant Rabbit Monoclonal Antibody (110)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA531196
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse LIFR.
Signal-transducing molecule. May have a common pathway with IL6ST. The soluble form inhibits the biological activity of LIF by blocking its binding to receptors on target cells.
Specifications
| LIFR | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P42703 | |
| Lifr | |
| Recombinant Mouse LIFR protein (Met1-Ser828). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA | |
| 110 | |
| Unconjugated | |
| Lifr | |
| A230075M04Rik; AW061234; CD118; CD118 antigen; D-factor/LIF receptor; FLJ98106; FLJ99923; leukemia inhibitor factor receptor alpha-chain; leukemia inhibitory factor receptor; leukemia inhibitory factor receptor alpha; leukemia inhibitory factor receptor alpha chain; LIF; LIF R beta; LIF receptor; Lifr; LIF-R; SJS2; soluble differentiation-stimulating factor receptor; STWS; SWS | |
| Rabbit | |
| Protein A | |
| RUO | |
| 16880 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.