Learn More
Abeba™ Light 1120 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand: Abeba™ 1120/35
Additional Details : Weight : 1.04000kg
Description
- CE EN ISO 20347:2012 O1 FO SRA
- Smooth leather, perforated
- White
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3559)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Double velcro fastening
- Reflective material
Specifications
CE EN ISO 20347:2012 O1 FO SRA | |
Leather | |
Slip-resistant | |
1120 |
White | |
35 | |
Polyurethane |