Learn More
Abeba™ Light 1141 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand: Abeba™ 1141/40
Additional Details : Weight : 0.80000kg
Description
- CE EN ISO 20347:2012 OB A, E, FO SRA
- Smooth leather white
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3558)
- Sanitized treated
- Antistatic
- Heel strap foldable, adjustable
- Breathable lining with Silver ions
- Instep area with elastic band
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRA | |
Leather | |
Slip-resistant | |
1141 |
White | |
40 | |
Polyurethane |