Learn More
Fisherbrand™ Mini Light Box
Product Code. 12852869 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Length (Metric):
161 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12852869 161 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12852869
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Store it in a drawer to save work space. Fisherbrand™ Mini Light Box is perfect for quick screening of well plates, Petri dishes, gels, slides and transparencies. The battery operated unit keeps illumination within arm's reach without cluttering a crowded benchtop.
  • Perfect for space-challenged labs or classrooms
  • Can be stored in a drawer or with limited footprint on the benchtop
  • 10 x 127mm viewing area is ideal for 96-well plates, Petri dishes, electrophoresis gels, slides and transparencies
  • Powered by four AA batteries (included) for a portable light box
Specifications
Battery Type AAA
For Use With (Equipment) 96-Well Plates, Petri Dishes, Electrophoresis Gels, Slides and Transparencies
Height (Metric) 44 mm
Length (Metric) 161 mm
Width (Metric) 135 mm