missing translation for 'onlineSavingsMsg'
Learn More

Light Green SF Yellowish (Technical Grade), TRC

Product Code. 30467925
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Product Code. Quantity unitSize
30467925 2.5 g 2.5g
1 options
This item is not returnable. View return policy

Product Code. 30467925

Brand: Toronto Research Chemicals TRCL3979682.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Light Green SF
CAS 5141-20-8
Molecular Formula C37 H34 N2 O9 S3 . 2 Na
Quantity 2.5 g
Synonym Benzenemethanaminium, N-ethyl-N-[4-[[4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl](4-sulfophenyl)methylene]-2,5-cyclohexadien-1-ylidene]-3-sulfo-, hydroxide, inner salt, disodium salt, Benzenemethanaminium, N-ethyl-N-[4-[[4-[ethyl[(3-sulfophenyl)methyl]ami
InChI Formula InChI=1S/C37H36N2O9S3.2Na/c1-3-38(25-27-7-5-9-35(23-27)50(43,44)45)32-17-11-29(12-18-32)37(31-15-21-34(22-16-31)49(40,41)42)30-13-19-33(20-14-30)39(4-2)26-28-8-6-10-36(24-28)51(46,47)48;;/h5-24H,3-4,25-26H2,1-2H3,(H2-,40,41,42,43,44,45,46,47,48);;/q;2*+1/
SMILES [Na+].[Na+].CCN(Cc1cccc(c1)S(=O)(=O)[O-])c2ccc(cc2)C(=C3C=CC(=[N+](CC)Cc4cccc(c4)S(=O)(=O)[O-])C=C3)c5ccc(cc5)S(=O)(=O)[O-]
IUPAC Name disodium;3-[[N-ethyl-4-[[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]-(4-sulfonatophenyl)methyl]anilino]methyl]benzenesulfonate
Molecular Weight (g/mol) 792.848
Formula Weight 792.122
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.