missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid Alexa Fluor 594 Antibody Labeling Kit

Product Code. 18098354
Change view
Click to view available options
Quantity:
1 mg
100 μg
30 μg
300 μg
Unit Size:
1 set
100µg
1mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18098354 30 μg 1 set
18051016 100 μg 100µg
18084126 300 μg 1 set
18093774 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18098354 Supplier Novus Biologicals™ Supplier No. 3350030
  • ONLINE EXCLUSIVE PRICE
    612.00€
    578.55€ / 1 sets
    Save 33.45€
    5% Off
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Antibody labeling kit

Lightning-Link™ Rapid Antibody Labeling kits enable direct labeling of antibodies, proteins, peptides or other biomolecules and allow conjugations to be set up in seconds, and used within minutes (less than 20 minutes).

Kit Contents:

  • Lightning-Link™ Rapid mix
  • 1 vial of LL Rapid Quencher reagent
  • 1 vial of LL Rapid Modifier reagent
TRUSTED_SUSTAINABILITY

Specifications

Quantity 30 μg
Product Type Antibody Labeling Kit
Content And Storage -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.