Learn More
Novus Biologicals™ Lightning-Link (R) Rapid DyLight 550 Antibody Labeling Kit
Product Code. 18014546 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18014546 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18014546 Supplier Novus Biologicals™ Supplier No. 3230005
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
Antibody labeling kit
Lightning-Link™ Rapid Antibody Labeling kits enable direct labeling of antibodies, proteins, peptides or other biomolecules and allow conjugations to be set up in seconds, and used within minutes (less than 20 minutes).
Kit Contents:
  • Lightning-Link™ Rapid mix
  • 1 vial of LL Rapid Quencher reagent
  • 1 vial of LL Rapid Modifier reagent
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 μg
Product Type Antibody Labeling Kit
Content And Storage -20°C