Learn More
Biochrom Lightwave 3
Product Code. 17143628
Change view
Click to view available options
Includes:
Spec, Bluetooth Connectivity
Spec, Bluetooth, Printer
Spec, Built-in Serial Printer
Spectrophotometer
Interface:
Large Disp,Button UI,Bluetooth
Large Display, Button UI
Printer Included:
No
Yes
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Interface Printer Included unitSize
17143628 Spectrophotometer Large Display, Button UI No Each
17253064 Spec, Built-in Serial Printer Large Display, Button UI Yes Each
17263064 Spec, Bluetooth Connectivity Large Disp,Button UI,Bluetooth No Each
17273064 Spec, Bluetooth, Printer Large Disp,Button UI,Bluetooth Yes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17143628 Supplier Biochrom Supplier No. 80300742
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
The Lightwave 3 is a split beam UV/Vis spectrophotometer that combines ease of use with the flexibility of a 40 mm sample port.
  • Split Beam (RBC) UV/Vis Spectrophotometer covering 190 to 1100 nm (5 nm bandwidth)
  • USB port for data collection
  • Accepts 10, 20, and 40 mm path length cuvettes
  • Built-in software provides flash scan, fixed wavelength measurements, kinetics, and concentration with comprehensive graphics capability plus the ability to store up to 90 methods
  • Choose to include a built-in serial printer, Bluetooth connectivity, or both
  • Lightweight and stackable for storage
Specifications
Beam Center Height 15 mm
Depth (English) 15.3 in.
Depth (Metric) 390 mm
Detector Type Twin CMOS Array
Display Graphic LCD
Languages English, French, German, Spanish, Italian, Chinese, Japanese
Light Source Xenon Flash
Product Line Lightwave 3
Spectral Bandwidth 5 nm
Type UV-Vis Spectrophotometer
Voltage 120/240 V
Wavelength Range 190 to 1100 nm
For Use With (Application) UV/Vis Spectrophotometry
Height (English) 3.9 in.
Height (Metric) 100 mm
Includes Spectrophotometer
Interface Large Display, Button UI
Monochromator Flat Grating
Noise ±0.005 peak to peak ±0.002 RMS
Photometric Linearity ±1.3% or ±0.008 A (Whichever is Greater at 546 nm)
Photometric Range -0.3 to 2.5 A, 0.3 to 199 %T
Photometric Repeatability ±0.002 A to 0.5 A (at 546 nm)
Printer Included No
Shape Round
Stray Light <0.5 %T 340 nm
Wavelength Accuracy ±2 nm
Wavelength Repeatability ±1 nm
Weight (English) 6.6 lb.
Weight (Metric) 3 kg
Width (English) 10 in.
Width (Metric) 260 mm