missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LILRB2 Recombinant Rabbit Monoclonal Antibody (9A3J8)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558210

 View more versions of this product

Product Code. 30282626

  • 436.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: QTGTIPKPTL WAEPDSVITQ GSPVTLSCQG SLEAQEYRLY REKKSASWIT RIRPELVKNG QFHIPSITWE HTGRYGCQYY SRARWSELSD PLVLVMTGAY PKPTLSAQPS PVVTSGGRVT LQCESQVAFG GFILCKEGED EHPQCLNSQP HARGSSRAIF SVGPVSPNRR WSHRCYGYDL NSPYVWSSPS DLLELLVPGV SKKPSLSVQP GPVMAPGESL TLQCVSDVGY DRFVLYKEGE RDLRQLPGRQ PQAGL.

CD85d, also known as LILRB2 or ILT4, is a member of the immunoglobulin-like transcript (ILT), leukocyte Ig-like receptor (LIR), and monocyte Ig-like receptor (MIR) family. It is a 110 kDa single transmembrane receptor with three Ig-like domains and a long cytoplasmic domain containing immunoreceptor tyrosine-based inhibitory motifs (ITIMs). CD85d is expressed on monocytes, macrophages, and dendritic cells. The receptor binds to non-classical class I molecules such as HLA-G, -B, and G1. Upon ligand binding, the tyrosine phosphatase SHP-1 interacts with the ITIMs, transducing a negative signal that inhibits immune response stimulation. This interaction is thought to contribute to tolerance during fetal implantation and plays a role in controlling inflammatory responses and cytotoxicity, helping to focus the immune response and limit autoreactivity. CD85d is also involved in organ allograft rejection, with the levels of ILT3 and ILT4 on dendritic cells suggesting its role in this process. The LILRB2 gene, located in a gene cluster at chromosomal region 19q13.4, encodes multiple transcript variants with different isoforms, highlighting its complexity and importance in immune regulation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LILRB2
Recombinant Monoclonal
0.25 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Q8N423
LILRB2
Recombinant fusion protein containing a sequence corresponding to amino acids 22-461 of human ILT4 (NP_001074447.2).
100 μL
Primary
Human
Antibody
IgG
ELISA, Immunocytochemistry
9A3J8
Unconjugated
LILRB2
CD85 antigen-like family member D; CD85D; CD85d antigen; Ig-like transcript 4; ILT4; ILT-4; leucocyte Ig-like receptor B2; leukocyte immunoglobulin like receptor B2; leukocyte immunoglobulin-like receptor subfamily B member 2; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 2; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 3; LILRB2; Lilrb3; LIR2; LIR-2; LIR2CD85D; MIR10; MIR-10; MIR10LILRA6; monocyte/macrophage immunoglobulin-like receptor 10; myeloid inhibitory receptor 10; Neutrophil immunoglobulin-like receptor 1; Nilr1; NILR-1; paired-Ig-like receptor B; Pirb; type 1 one-pass transmembrane Ig-like receptor
Rabbit
Protein A
RUO
10288
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.