missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LILRB5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12881-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene is a member of the leukocyte immunoglobulin-likereceptor (LIR) family, which is found in a gene cluster atchromosomal region 19q13.4. The encoded protein belongs to thesubfamily B class of LIR receptors which contain two or fourextracellular immunoglobulin domains, a transmembrane domain, andtwo to four cytoplasmic immunoreceptor tyrosine-based inhibitorymotifs (ITIMs). Several other LIR subfamily B receptors areexpressed on immune cells where they bind to MHC class I moleculeson antigen-presenting cells and inhibit stimulation of an immuneresponse. Multiple transcript variants encoding different isoformshave been found for this gene.Specifications
| LILRB5 | |
| Polyclonal | |
| Unconjugated | |
| LILRB5 | |
| CD85C, LILRB5, LIR 8, LIR8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10990 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75023 | |
| LILRB5 | |
| LILRB5 Fusion Protein Ag3886 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction