missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Limitin/IFN-zeta Antibody (RM0123-7D19), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-12426CL1
This item is not returnable.
View return policy
Description
Limitin/IFN-zeta Monoclonal antibody specifically detects Limitin/IFN-zeta in Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Neutralization
Specifications
| Limitin/IFN-zeta | |
| Monoclonal | |
| CoraFluor 1 | |
| 6030405N23Rik, BGIF, Ifnz, interferon zeta, Lmtn | |
| Mouse recombinant Limitin/IFN-zeta | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Neutralization | |
| RM0123-7D19 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 319146 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction