Learn More
LIMK1 Monoclonal Antibody (OTI3G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502975
Description
There are approximately 40 known eukaryotic LIM proteins, so named for the LIM domains they contain. LIM domains are highly conserved cysteine-rich structures containing 2 zinc fingers. Although zinc fingers usually function by binding to DNA or RNA, the LIM motif probably mediates protein-protein interactions. LIM kinase-1 and LIM kinase-2 belong to a small subfamily with a unique combination of 2 N-terminal LIM motifs and a C-terminal protein kinase domain. LIMK1 is likely to be a component of an intracellular signaling pathway and may be involved in brain development. LIMK1 hemizygosity is implicated in the impaired visuospatial constructive cognition of Williams syndrome.Specifications
| LIMK1 | |
| Monoclonal | |
| Unconjugated | |
| LIMK1 | |
| LIMK, LIMK-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3984 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3G3 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P53667 | |
| LIMK1 | |
| Full length human recombit protein of human LIMK1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.