missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ROBO2 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB3147T-100UG
This item is not returnable.
View return policy
Description
ROBO2 Monoclonal specifically detects ROBO2 in Human samples. It is validated for Flow Cytometry.
Specifications
| ROBO2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9HCK4.2 | |
| ROBO2 | |
| Mouse myeloma cell line NS0-derived recombinant human ROBO2 isoform 1 Ser22-Pro859 Accession # Q9HCK4.2 | |
| RUO | |
| 6092 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 356001 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Drosophila) homolog 2, roundabout, axon guidance receptor, homolog 2 (Drosophila), SAX3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human ROBO2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ROBO1, rhROBO3, or rhROBO4 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction