Learn More
Lipin 1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB110-57150PCP
Product Code. 18543050
  • 558.00€ / 0.10mL
Explore more special offers
Description
Description
Lipin 1 Polyclonal antibody specifically detects Lipin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)