Learn More
Hamilton™ Liq-Glass™ 400 Electrodes
Product Code. 10225889
Change view
Click to view available options
Connector Types:
Electrical S7
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
10225889 Electrical S7 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10225889 Supplier Hamilton™ Supplier No. 238183
  • ONLINE EXCLUSIVE PRICE
    354.26€
    327.08€ / Pack of 1
    Save 27.18€
    7% Off
Estimated Shipment: 26-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Order Info
FillingSolutionType
Specifications
Connector Types Electrical S7
Material Glass
Internal Reference Hamilton Everef
Length (Metric) 400 mm
Diameter (Metric) Body 12 mm