missing translation for 'onlineSavingsMsg'
Learn More

Lithium aluminum hydride, 95% min, Thermo Scientific™

Reducing Agent For Pharmaceuticals, Perfumes, and Fine organic Chemicals; Source of Hydrogen

Brand:  Thermo Scientific Alfa Aesar 013442.51

Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11326788

  • ONLINE EXCLUSIVE PRICE
    168.00€
    164.99€ / 125g
    Save 3.01€
    1% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

16853-85-3
95% min.
MFCD00011075
Decomposes in water. Soluble in ether,diethyl ether and terahydrofuran.
[Li+].[AlH4-]
37.95
37.95
Air and moisture sensitive
0.917
5 x 25g
AlH4Li
lithium aluminum hydride, lithium aluminium hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
OCZDCIYGECBNKL-UHFFFAOYSA-N
lithium(1+) alumanuide
21226445
Odorless
Light gray powder
Lithium aluminum hydride
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.