missing translation for 'onlineSavingsMsg'
Learn More

Lithium aluminum hydride, 95% min, Thermo Scientific™

Reducing Agent For Pharmaceuticals, Perfumes, and Fine organic Chemicals; Source of Hydrogen

Brand:  Thermo Scientific Alfa Aesar 013442.22

Quantity 100g

 View more versions of this product

Product Code. 11346788

  • ONLINE EXCLUSIVE PRICE
    140.00€
    137.00€ / 100g
    Save 3.00€
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reducing agent for pharmaceuticals, perfumes and fine organic chemicals; source of hydrogen, propellant, and catalyst in polymerizationsLithium aluminum hydride is used as a reducing agent in various synthetic organic chemistry reactions. For example, it is used in the conversion of esters, carboxylic acids, acyl chlorides, aldehydes and ketones into their corresponding alcohols. It is also involved in the conversion of amide, nitro, nitrile, imine, oxime and azide compounds into their amines as well as reduces alkyl halides to alkanes. It reacts with sodium chloride to prepare sodium hydride. It acts as a potential hydrogen storage medium, which could be used for future fuel cell-powered vehicles. It is mainly used in the preparation of other tetrahydridoaluminumates like sodium aluminum hydride, potassium aluminum hydride and magnesium alanate. Furthermore, it has been used to prepare thermoplastic polyester polyamides from oleic acid and lithium-polymer batteries.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

16853-85-3
37.95
OCZDCIYGECBNKL-UHFFFAOYSA-N
21226445
[Li+].[AlH4-]
AlH4Li
MFCD00011075
lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
lithium(1+) alumanuide

Specifications

16853-85-3
AlH4Li
UN1410
lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
OCZDCIYGECBNKL-UHFFFAOYSA-N
lithium(1+) alumanuide
21226445
≥95%
Air and moisture sensitive
Lithium aluminum hydride
100g
MFCD00011075
14,349
Decomposes in water; Soluble in ether,diethyl ether and terahydrofuran.
[Li+].[AlH4-]
37.95
37.95
Odorless
0.917
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.