Learn More
Lithium Fluoride, Honeywell Fluka™
Product Code. 15690540 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15690540 1 kg 1kg
15680540 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15690540 Supplier Honeywell-Fluka Supplier No. 011401KG
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Purum,≥99%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7789-24-4
Molecular Formula FLi
Molecular Weight (g/mol) 25.94
MDL Number MFCD00011090
InChI Key PQXKHYXIUOZZFA-UHFFFAOYSA-M
Synonym lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
PubChem CID 224478
IUPAC Name lithium;fluoride
SMILES [Li+].[F-]
Specifications
Quantity 1 kg
Linear Formula LiF
UN Number 3288
Formula Weight 25.94g/mol
Percent Purity ≥99%
Grade purum
Density 2.64g/mL at 25°C
Chemical Name or Material Lithium Fluoride