missing translation for 'onlineSavingsMsg'
Learn More

Lithium Fluoride, TRC

Product Code. 30469815
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30469815 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30469815 Supplier Toronto Research Chemicals Supplier No. TRCL3976985G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium fluoride
CAS 7789-24-4
Quantity 5 g
Molecular Formula LiF
InChI Formula InChI=1S/FH.Li/h1H;/q;+1/p-1
SMILES [Li+].[F-]
IUPAC Name lithium;fluoride
Molecular Weight (g/mol) 25.938
Formula Weight 26.014
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.