Learn More
BD Vacutainer Lithium Heparin Tubes
Product Code. 15753036
Change view
Click to view available options
Capacity (Metric):
4 mL
6 mL
Tube Size:
13 x 75 mm
13 x 100 mm
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Tube Size unitSize
15753036 4 mL 13 x 75 mm Pack of 1000
12834811 6 mL 13 x 100 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15753036 Supplier BD Vacutainer Supplier No. 368884
Plasma tube for clinical chemistry includes spray-dried lithium heparin as an anticoagulant.
  • Plasma tubes for clinical chemistry with spray-dried lithium heparin.
  • Heparin acts as an anticoagulant as it develops an antithrombin complex.
  • Plasma tubes should be gently inverted 180° and back 8-10 times.
  • Correct mixing immediately after the blood sample has been taken is extremely important to avoid microclotting.
  • The lithium heparin is spray dried onto the inner walls of the tubes using a special procedure so that the additive is evenly distributed to achieve the best possible solubility.
  • For clinical chemistry, lithium heparin is generally preferred over sodium heparin.
TRUSTED_SUSTAINABILITY
Specifications
Additive Lithium Heparin
Material PET
Capacity (Metric) 4 mL
Closure Color Green
Tube Size 13 x 75 mm
Label Type Paper