missing translation for 'onlineSavingsMsg'
Learn More

Lithium Hydroxide Monohydrate, Certified AR for Analysis, Fisher Chemical

Product Code. 10101530 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10101530 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10101530 Supplier Fisher Chemical Supplier No. L/2301/53
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 920°C
Melting Point 462°C
CAS 1310-66-3
Color White
pH 12
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula H3LiO2
MDL Number MFCD00149772
Synonym lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
SMILES [Li+].O.[OH-]
IUPAC Name lithium(1+) hydrate hydroxide
Molecular Weight (g/mol) 41.96
PubChem CID 168937
Formula Weight 41.95g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.