missing translation for 'onlineSavingsMsg'
Learn More

Lithium Perchlorate Trihydrate, Honeywell Fluka™

Product Code. p-7120523 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 15651760

Brand: Honeywell-Fluka 20530350G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

95-102 %

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13453-78-6
Molecular Formula ClH6LiO7
Molecular Weight (g/mol) 160.431
MDL Number MFCD00149765
InChI Key JAWYRNYHJJDXHX-UHFFFAOYSA-M
Synonym lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion
PubChem CID 23708903
IUPAC Name lithium;perchlorate;trihydrate
SMILES [Li+].O.O.O.[O-]Cl(=O)(=O)=O

Specifications

Color White
Quantity 50 g
Linear Formula LiClO4 · 3H2O
UN Number UN1481
Formula Weight 160.44g/mol
Packaging Poly Bottle
Appearance Crystals
Density 1.841g/mL at 25°C
Chemical Name or Material Lithium Perchlorate Trihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.