missing translation for 'onlineSavingsMsg'
Learn More

Lithium perchlorate trihydrate, Reagent Grade

Product Code. 11381738
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 11381738

Brand: Thermo Scientific Alfa Aesar 011638.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13453-78-6 | ClH6LiO7 | 160.431 g/mol

It is used as an oxygen source for chemical oxygen generators, as it liberates oxygen at high temperatures. Excellent solubility of lithium perchlorate permits its use in Diels-Alder reactions. It is also employed in lithium-ion batteries as an electrolyte, and act as chaotropic agent to denature proteins. It is used in Baylis-Hillman reaction as a catalyst in the coupling of alpha,beta-unsaturated carbonyl compounds with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 13453-78-6
Molecular Formula ClH6LiO7
Molecular Weight (g/mol) 160.431
MDL Number MFCD00149765
InChI Key JAWYRNYHJJDXHX-UHFFFAOYSA-M
Synonym lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion
PubChem CID 23708903
IUPAC Name lithium;perchlorate;trihydrate
SMILES [Li+].O.O.O.[O-]Cl(=O)(=O)=O

Especificaciones

Color White
Physical Form Crystalline
Quantity 1 kg
Linear Formula LiClO4·3H2O
UN Number UN1481
Merck Index 14,5539
Solubility Information Highly soluble in water,methanol,ethanol,ethyl acetate,diethyl ether,and acetone.
Formula Weight 160.45 (106.40 Anhydrous)
Grade Reagent
Odor Odorless
Density 1.84 g/mL
Chemical Name or Material Lithium perchlorate trihydrate
Mostrar más Mostrar menos

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado