missing translation for 'onlineSavingsMsg'
Learn More

Lithium Tetrafluoroborate, TRC

Product Code. 30475227
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475227 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30475227 Supplier Toronto Research Chemicals Supplier No. TRCL46915050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium Tetrafluoroborate
CAS 14283-07-9
Molecular Formula BF4Li
Quantity 50 g
Synonym Lithium Fluoborate (6CI), Lithium Tetrafluoroborate (7CI), Lithium Boridefluoride (LiBF4), Lithium Fluoroborate, Lithium Fluoroborate (LiBF4), Lithium tetrafluoroborate (LiBF4), Lithium tetrafluoroborate(1-)
InChI Formula InChI=1S/BF4.Li/c2-1(3,4)5;/q-1;+1
SMILES [Li+].F[B-](F)(F)F
IUPAC Name lithium;tetrafluoroborate
Molecular Weight (g/mol) 93.75
Formula Weight 94.0189
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.