missing translation for 'onlineSavingsMsg'
Learn More

Lithium Trifluoromethylsulfonate, TRC

Product Code. 30477163
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477163 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477163 Supplier Toronto Research Chemicals Supplier No. TRCL46917010G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium Trifluoromethylsulfonate
CAS 33454-82-9
Molecular Formula CF3LiO3S
Quantity 10 g
Synonym Trifluoromethanesulfonic Acid Lithium Salt, AQ 75T, EF 15, Fluorad FC 122, LiTFS, Lithium Triflate, PFM-Li, Sankonol AQ 20T, Sankonol AQ 50T, Sankonol AQ 75T, Sankonol EAc 30T
InChI Formula InChI=1S/CHF3O3S.Li/c2-1(3,4)8(5,6)7;/h(H,5,6,7);/q;+1/p-1
SMILES [Li+].[O-]S(=O)(=O)C(F)(F)F
IUPAC Name lithium;trifluoromethanesulfonate
Molecular Weight (g/mol) 156.01
Formula Weight 155.968
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.