missing translation for 'onlineSavingsMsg'
Learn More

Litmus, MP Biomedicals™

Product Code. 11493462 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
Unit Size:
25g

Product Code. 11493462

Brand: MP Biomedicals 0215526325

Please to purchase this item. Need a web account? Register with us today!

Litmus, Dark blue powder

  • An acid-base indicator developed from various orcinol-containing lichen including Lecanora tartarea, Rocella fuciformis and Variolaria dealbata.
  • The principal coloring matter in Litmus is Azolitmus, a substituted 7-hydroxy-2-phenoxazone derivative.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1393-92-6
MDL Number MFCD00131528
Synonym C.I. Natural Red 28, Lackmus, Tournesole, Chhadila

Specifications

Color Dark Blue
pH 5 to 8 (literature)
Quantity 25 g
Solubility Information Soluble in ethanol (3mg/mL); partly soluble water (10mg/mL-opaque dark blue solution) or alcohol.
Physical Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.