missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Livin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97074-100ul
This item is not returnable.
View return policy
Description
Livin Polyclonal antibody specifically detects Livin in Human samples. It is validated for ELISA
Specifications
| Livin | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| baculoviral IAP repeat containing 7, baculoviral IAP repeat-containing 7, KIAPRING finger protein 50, Kidney inhibitor of apoptosis protein, Livin, livin inhibitor-of-apoptosis, Melanoma inhibitor of apoptosis protein, MLIAPlivin inhibitor of apoptosis, ML-IAPmliap, RNF50LIVINbaculoviral IAP repeat-containing protein 7 | |
| Produced in rabbits immunized with purified, recombinant Human Livin (Accession#: AAH14475.1; Met1-Ser298) | |
| 100 μg | |
| Apoptosis, Zinc Finger | |
| 79444 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction