Learn More
LMNA Monoclonal Antibody (OTI2G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA803495S
Encompass_Preferred
 View more versions of this product
Product Code. 17532623
  • 242.00€ / 30µL
Description
LMNA Monoclonal antibody specifically detects LMNA in Human samples. It is validated for Western Blot
Specifications
Specifications
LMNA
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P02545
Mouse
Affinity chromatography
RUO
4000
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI2G1
Unconjugated
LMNA
70 kDa lamin; CDCD1; CDDC; CMD1A; CMT2B1; Dhe; EMD2; FPL; FPLD; FPLD2; HGPS; IDC; lamin; lamin A; lamin A/C; lamin A/C-like 1; lamin C; lamin C2; lamin-A/C; LDP1; LFP; LGMD1B; LMN1; Lmna; LMNA protein; LMNC; LMNL1; OTTHUMP00000015843; OTTHUMP00000015846; OTTHUMP00000015847; Prelamin-A/C; PRO1; renal carcinoma antigen NY-REN-32; unnamed protein product
Full length human recombinant protein of human LMNA produced in HEK293T cell.