Learn More
Heinz Herenz LMS-Swab, Amies
Click to view available options
Tip Type:
Fine
Standard
Unit Size:
Pack of 100
Pack of 900
Description
- For transportation and preservation of microbiological samples
- Liquid media allows for easy dilution of samples
- Carry out differnt tests out of one sample
- Ready-to-use device delivered in an easy-to-open peel pouch
- Breakpoint in the shaft if the applicator allows for a safe and hygienic work-flow
- Pre-labeled test tubes allow for correct identification
- Produced according to GMP (Good Manufacturing Practice) and certified according to DIN EN ISO 13485 ensure the high quality
- Sterile (SAL 10-6)
Specifications
Specifications
| Product Type | Transport and Collection Swab |
| Sterility | Sterile |
| Closure Color | Pink |
| Description | Pyrogen-free and non-cytotoxic |
| Includes | Test tube with screw cap and liquid media |
| Packaging | Peel pouch |
| Shaft Material | Foam stick |
| Tip Type | Fine |