Learn More
- Home
- Products
- Thermometers and Temperature Measurement
- Manual Thermometers
- Brannan™ LO-tox™ Lab Thermometer with 3-points Traceable Calibration
Brannan™ LO-tox™ Lab Thermometer with 3-points Traceable Calibration
Product Code. p-200007110
Change view
Click to view available options
Temperature Range (Metric):
-1°C to 101°C
-1°C to 51°C
-10°C to +110°C
-10°C to +210°C
-10°C to +260°C
-10°C to +300°C
-10°C to +330°C
-10°C to +360°C
-10°C to +50°C
-20°C to +110°C
-20°C to +150°C
-35°C to +50°C
Subdivision:
0.1°C
0.2°C
0.5°C
1°C
2°C
Immersion Type:
Partial Immersion
Total Immersion
Length (Metric):
15.5 cm
30.5 cm
40.5 cm
Unit Size:
Each
24 product options available for selection
| Product Code. | Temperature Range (Metric) | Subdivision | Immersion Type | Length (Metric) | Unit Size |
|---|---|---|---|---|---|
| 17823008 | -10°C to +330°C | 2°C | Partial Immersion | 30.5 cm | Each |
| 17873028 | -20°C to +110°C | 0.5°C | Partial Immersion | 30.5 cm | Each |
| 17862988 | -1°C to 101°C | 0.2°C | Partial Immersion | 40.5 cm | Each |
| 17833028 | -10°C to +50°C | 0.5°C | Total Immersion | 30.5 cm | Each |
| 17883028 | -10°C to +260°C | 1°C | Partial Immersion | 40.5 cm | Each |
| 17822998 | -10°C to +110°C | 1°C | Total Immersion | 15.5 cm | Each |
| 17842998 | -10°C to +360°C | 2°C | Total Immersion | 30.5 cm | Each |
| 17863018 | -1°C to 51°C | 0.1°C | Partial Immersion | 40.5 cm | Each |
| 17843028 | -10°C to +210°C | 1°C | Partial Immersion | 40.5 cm | Each |
| 17892988 | -1°C to 101°C | 0.2°C | Total Immersion | 40.5 cm | Each |
| 17853018 | -35°C to +50°C | 1°C | Partial Immersion | 30.5 cm | Each |
| 17843038 | -20°C to +110°C | 0.5°C | Total Immersion | 30.5 cm | Each |
| 17823068 | -1°C to 51°C | 0.2°C | Total Immersion | 40.5 cm | Each |
| 17803088 | -10°C to +300°C | 1°C | Partial Immersion | 30.5 cm | Each |
| 17853108 | -20°C to +110°C | 1°C | Partial Immersion | 30.5 cm | Each |
| 17893118 | -10°C to +210°C | 1°C | Total Immersion | 40.5 cm | Each |
| 17813088 | -20°C to +150°C | 1°C | Partial Immersion | 30.5 cm | Each |
| 17873108 | -20°C to +110°C | 1°C | Total Immersion | 30.5 cm | Each |
| 17893108 | -1°C to 51°C | 0.2°C | Partial Immersion | 40.5 cm | Each |
| 17853088 | -20°C to +150°C | 1°C | Total Immersion | 30.5 cm | Each |
| 17833108 | -10°C to +50°C | 0.5°C | Partial Immersion | 30.5 cm | Each |
| 17863068 | -1°C to 51°C | 0.1°C | Total Immersion | 40.5 cm | Each |
| 17833078 | -10°C to +50°C | 1°C | Total Immersion | 15.5 cm | Each |
| 17823078 | -35°C to +50°C | 0.5°C | Total Immersion | 30.5 cm | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
This item is not returnable.
View return policy
Product Code.
17823008
Supplier
Brannan™
Supplier No.
44/814/0 + 49/101/0
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable.
View return policy
Description
- LO-tox™ is an organic filling specially chosen to achieve a wide temperature span and provide the lowest practical levels of hazard
- Safe non-mercury filling
- Supplied with calibration certificate
- Non-roll end cap
- Supplied as standard in protective individual, resealable telescopic tube
- Made in UK
Specifications
Specifications
| Calibration Points | 0°C, 50°C, 90°C |
| Certifications/Compliance | UK Traceable to National Standards ISO 9001 |
| Immersion Type | Partial Immersion |
| Length (Metric) | 30.5 cm |
| Material | Glass |
| Thermometer Fill | Brown Spirit |
| Max. Temperature (English) | 626°F |
| Max. Temperature (Metric) | +330°C |
| Min. Temperature (English) | 14°F |
| Min. Temperature (Metric) | -10°C |
| Temperature Range (English) | 14°F to 626°F |
| Temperature Range (Metric) | -10°C to +330°C |
| Accuracy | ± 2.0°C at 90°C |
| Subdivision | 2°C |
| Immersion Length | 76 mm |
| Width (Metric) | 1.2 cm |
| Show More Show Less |