Learn More
Notrax Logo Standard™ Affordable Absorbent Printed Logo Mat
Product Code. 17722018
Change view
Click to view available options
Dimensions (L x W):
90 x 60 cm
120 x 90 cm
150 x 90 cm
300 x 90 cm
180 x 120 cm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
17722018 90 x 60 cm Each
17732018 120 x 90 cm Each
17742018 150 x 90 cm Each
17752018 180 x 120 cm Each
17762018 300 x 90 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17722018 Supplier Notrax Supplier No. 193S0023WH
  • ONLINE EXCLUSIVE PRICE
    53.55€
    46.20€ / Pack of 1
    Save 7.35€
    13% Off
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
Logo Standard™ can be fully customized with BrilliantStep™ state-of-the-art printing technology allowing 3-dimensional effects, fine lines and color shading, for each mat to have its truly individual image.
  • Logo Standard™ is the ideal promotional matting solution for light duty applications
  • Using BrilliantStep™ full color printing with almost infinite color combinations, the even velour surface allows fine printing and true color presentation
  • A durable vinyl backing with smooth finish adapts to any surface
  • Logo Standard™ can be fully customized with BrilliantStep™ state-of-the-art printing technology allowing 3-dimensional effects, fine lines and color shading, for each mat to have its truly individual image
Specifications
Dimensions (L x W) 90 x 60 cm
Style Frieze
Color Logo
Length (English) 35 in.
Length (Metric) 90 cm
Material Polyamide 6.6 High Twist Yarns
Type Industrial and Commercial Matting
Width (Metric) 60 cm