missing translation for 'onlineSavingsMsg'
Learn More

LONP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 11 publications

369.00€ - 557.00€

Specifications

Antigen LONP1
Concentration 0.2mg/mL
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18418820
View Documents
Novus Biologicals
NBP1-81734-25ul
25 μL
369.00€
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18765153
View Documents
Novus Biologicals
NBP1-81734
0.1 mL
557.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LONP1 Polyclonal specifically detects LONP1 in Human, Mouse, Rat, Drosophila samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications

Specifications

LONP1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat, Drosophila
EC 3.4.21, EC 3.4.21.-, EC 3.4.21.53, hLON, hLON ATP-dependent protease, LON, lon peptidase 1, mitochondrial, lon protease homolog, mitochondrial, Lon protease-like protein, LONHs, LONP, Mitochondrial ATP-dependent protease Lon, mitochondrial lon peptidase 1, mitochondrial lon protease-like protein, PIM1, protease, serine, 15, PRSS15MGC1498, Serine protease 15
LONP1
IgG
Affinity Purified
Specificity of human LONP1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
0.2mg/mL
Polyclonal
Rabbit
Cancer, Epigenetics, metabolism
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
9361
This antibody was developed against Recombinant Protein corresponding to amino acids:VEEKIKQTHRKYLLQEQLKIIKKELGLEKDDKDAIEEKFRERLKELVVPKHVMDVVDEELSKLGLLDNHSSEFNVTRNYLDWLTSIPWGKYSNENLDLARAQAVLEEDHYGMEDVKKRILEFIAVSQLRGSTQGKILCFYGP
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.