missing translation for 'onlineSavingsMsg'
Learn More

Losartan Trityl Ether, TRC

Product Code. 30138712
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138712 50 mg 50mg
17903295 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138712 Supplier Toronto Research Chemicals Supplier No. TRCL47053050MG
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1006062-28-7
Molecular Formula C41 H37 Cl N6 O
Molecular Weight (g/mol) 665.23
Synonym 2H-Tetrazole, 5-[4'-[[2-butyl-4-chloro-5-[(triphenylmethoxy)methyl]-1H-imidazol-1-yl]methyl][1,1'-biphenyl]-2-yl]- (ACI), 5-[4'-[[2-Butyl-4-chloro-5-[(triphenylmethoxy)methyl]-1H-imidazol-1-yl]methyl][1,1'-biphenyl]-2-yl]-2H-tetrazole (ACI), 5-[4'-[[2-Butyl-4-chloro-5-[[(triphenylmethyl]oxy]methyl]-1H-imidazol-1-yl]methyl]biphenyl-2-yl]-1H-tetrazole, Losartan Potassium Imp. I (EP)

Specifications

Analyte or Component Names 5-[4'-[(2-Butyl-4-chloro-5-[((triphenylmethyl)oxy)methyl]-1H-imidazol-1-yl)methyl]biphenyl-2-yl]-1H-tetrazole
Shipping Condition Room Temperature
InChI Formula InChI=1S/C41H37ClN6O/c1-2-3-23-38-43-39(42)37(48(38)28-30-24-26-31(27-25-30)35-21-13-14-22-36(35)40-44-46-47-45-40)29-49-41(32-15-7-4-8-16-32,33-17-9-5-10-18-33)34-19-11-6-12-20-34/h4-22,24-27H,2-3,23,28-29H2,1H3,(H,44,45,46,47)
SMILES CCCCc1nc(Cl)c(COC(c2ccccc2)(c3ccccc3)c4ccccc4)n1Cc5ccc(cc5)c6ccccc6c7nnn[nH]7
IUPAC Name 5-[2-[4-[[2-butyl-4-chloro-5-(trityloxymethyl)imidazol-1-yl]methyl]phenyl]phenyl]-1H-tetrazole
Industry Type Pharmaceutical
Quantity 50 mg
Formula Weight 664.2717
Chemical Name or Material 5-[4'-[(2-Butyl-4-chloro-5-[((triphenylmethyl)oxy)methyl]-1H-imidazol-1-yl)methyl]biphenyl-2-yl]-1H-tetrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.