Learn More
LOX-1/OLR1 Antibody (331212), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB1798MFV610
Product Code. 18587356
  • 718.00€ / 0.10mL
Description
LOX-1/OLR1 Monoclonal antibody specifically detects LOX-1/OLR1 in Human samples. It is validated for Western Blot, Flow Cytometry, Blocking Assay, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LOX-1/OLR1
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Mouse
0.1 mL
Cardiovascular Biology, metabolism
4973
Store at 4C in the dark.
Western Blot, Flow Cytometry, Blocking Assay, CyTOF
331212
Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready
CLEC8ASLOX1, C-type lectin domain family 8 member A, hLOX-1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, Lectin-type oxidized LDL receptor 1, LOX-1, LOX1ox LDL receptor 1, LOXIN, oxidised low density lipoprotein (lectin-like) receptor 1, oxidized low density lipoprotein (lectin-like) receptor 1, oxidized low-density lipoprotein receptor 1, oxidized low-density lipoprotein receptor 1, soluble form, Ox-LDL receptor 1, SCARE1, scavenger receptor class E, member 1
Mouse myeloma cell line NS0-derived recombinant human LOX-1/OLR1, Ser61-Gln273, Accession # P78380
RUO
Primary
Human
IgG2b