missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Loxoprofen Sodium, Mikromol™
Description
Loxoprofen Sodium
Specifications
Specifications
| Chemical Name or Material | rac-Loxoprofen sodium salt |
| CAS | 80382-23-6 |
| Analyte or Component Names | rac-Loxoprofen sodium salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H17 O3 . Na |
| Certifications/Compliance | ISO 17025 |
| Synonym | α-Methyl-4-[(2-oxocyclopentyl)methyl]benzeneacetic Acid Sodium Salt, CS 600, Loxonin, Sodium 2-[4-[(2-Oxocyclopentyl)methyl]phenyl]propionate, Sodium Loxoprofen |
| InChI Formula | InChI=1S/C15H18O3.Na/c1-10(15(17)18)12-7-5-11(6-8-12)9-13-3-2-4-14(13)16;/h5-8,10,13H,2-4,9H2,1H3,(H,17,18);/q;+1/p-1 |
| SMILES | [Na+].CC(C(=O)[O-])c1ccc(CC2CCCC2=O)cc1 |
| IUPAC Name | sodium;2-[4-[(2-oxocyclopentyl)methyl]phenyl]propanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?