missing translation for 'onlineSavingsMsg'
Learn More

Loxoprofen Sodium (Mixture of diastereomers), TRC

Product Code. 30477424
Change view
Click to view available options
Quantity:
25 mg
5 g
Unit Size:
25mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30477424 5 g 5g
30467342 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477424 Supplier Toronto Research Chemicals Supplier No. TRCL4729005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-Loxoprofen sodium salt
CAS 80382-23-6
Molecular Formula C15 H17 O3 . Na
Quantity 5 g
Synonym Benzeneacetic acid, α-methyl-4-[(2-oxocyclopentyl)methyl]-, sodium salt (9CI), CS 600, CS 600 (antiinflammatory), Lobu, Loxonin, Loxoprofen sodium, Sodium 2-[4-[(2-Oxocyclopentyl)methyl]phenyl]propionate, Sodium loxoprofen
InChI Formula InChI=1S/C15H18O3.Na/c1-10(15(17)18)12-7-5-11(6-8-12)9-13-3-2-4-14(13)16;/h5-8,10,13H,2-4,9H2,1H3,(H,17,18);/q;+1/p-1
SMILES [Na+].CC(C(=O)[O-])c1ccc(CC2CCCC2=O)cc1
IUPAC Name sodium;2-[4-[(2-oxocyclopentyl)methyl]phenyl]propanoate
Molecular Weight (g/mol) 268.28
Formula Weight 268.1075
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.