Learn More
LPAR1/LPA1/EDG-2 Antibody (OTI1G6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-01354
Product Code. 18761364
  • 558.00€ / 0.10mL
Description
LPAR1/LPA1/EDG-2 Monoclonal specifically detects LPAR1/LPA1/EDG-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at -20°C. Avoid freeze-thaw cycles.
IgG2b