Learn More
LPAR1/LPA1/EDG-2 Antibody (SR1651), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21799-100ul
This item is not returnable.
View return policy
Description
LPAR1/LPA1/EDG-2 Monoclonal antibody specifically detects LPAR1/LPA1/EDG-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
Specifications
| LPAR1/LPA1/EDG-2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| SR1651 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
| edg-2, EDG2Lysophosphatidic acid receptor Edg-2, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor2,, Gpcr26, GPR26, LPA receptor 1, LPA-1, LPA1vzg-1, lysophosphatidic acid receptor 1, Mrec1.3, rec.1.3, ventricular zone gene 1, VZG1 | |
| A synthesized peptide derived from human LPAR1/LPA1/EDG-2 (Uniprot #: Q92633) | |
| 100 μg | |
| Apoptosis, GPCR, Signal Transduction | |
| 1902 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |