Learn More
DiscoverX LPI (soy), L-a-Lysophosphatidylinositol
Product Code. 17362449
Encompass
Change view
Click to view available options
:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17362449 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17362449 Supplier DiscoverX Supplier No. 921120
LPI (soy), L-a-Lysophosphatidylinositol is an agonist for GPR55. Provided as 1mg of dry powder.
L-α-Lysophosphatidylinositol (LPI) is an endogenous phospholipid and endocannabinoid neurotransmitter. This version is derived from soy. LPI is a novel ligand for the G protein-coupled receptor 55 (GPR55), for which it acts as an agonist. Also thought to be a promoter of RhoA-dependent calcium signaling, and an NFAT activator in embryonic kidney 293 cells. Provided as 1mg of dry powder.
Specifications
Quantity 1 mg