Learn More
LPIN1 Monoclonal Antibody (OTI1B2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA806208S
Encompass_Preferred
 View more versions of this product
Product Code. 17506303
  • 242.00€ / 30µL
Description
LPIN1 Monoclonal antibody specifically detects LPIN1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
LPIN1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P04150
Mouse
Affinity chromatography
RUO
2908
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI1B2
Unconjugated
Nr3c1
4631420P06; DKFZp781P1796; Fatty liver dystrophy protein; Fld; KIAA0188; lipin 1; Lipin1; lipin-1; LPIN1; mKIAA0188; PAP1; Phosphatidate phosphatase LPIN1
Human recombinant protein fragment corresponding to amino acids 1-265 of human NR3C1 produced in E.coli.