missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRIG2 Monoclonal Antibody (990332)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561071
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
| LRIG2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O94898 | |
| Lrig2 | |
| Chinese hamster ovary cell line CHO-derived recombinant human LRIG-2, Gly41-Thr805. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 990332 | |
| Unconjugated | |
| Lrig2 | |
| 4632419I10Rik; BB096938; Kiaa0806; leucine rich repeats and immunoglobulin like domains 2; leucine-rich repeats and immunoglobulin like domains 2; leucine-rich repeats and immunoglobulin-like domains 2; leucine-rich repeats and immunoglobulin-like domains protein 2; LIG2; LIG-2; Lrig2; UFS2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 9860 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction